Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen NDRG1
Applications Immunofluorescence, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093055
View Documents
Novus Biologicals
NBP303516100UL
100 μg Each for $320.71
Add to cart
 
NB093056
View Documents
Novus Biologicals
NBP30351620UL
20 μg Each for $138.37
Add to cart
 
Description

Description

NDRG1 Polyclonal antibody specifically detects NDRG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, KnockDown
Specifications

Specifications

NDRG1
Unconjugated
Rabbit
Cancer, Phospho Specific
PBS with 50% glycerol, pH7.3.
10397
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
CAP43, CMT4D, Differentiation-related gene 1 protein, DRG-1, DRG1HMSNL, NDR1, Nickel-specific induction protein Cap43, NMSL, N-myc downstream regulated 1, N-myc downstream-regulated gene 1 protein, protein NDRG1, protein regulated by oxygen-1, PROXY1, Reducing agents and tunicamycin-responsive protein, RIT42, RTPGC4, TARG1, TDD5, tunicamycin-responsive protein
Recombinant fusion protein containing a sequence corresponding to amino acids 175-394 of human NDRG1 (NP_001128714.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit