Learn More
Aviva Systems Biology Corp NDST4 Antibody - middle region (ARP49738_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49738_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDST4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NDST4. Alternate names: NDST-4, NHSST4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60164