Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDST4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548953
Description
Essential bifunctional enzyme that catalyzes both the N- deacetylation and the N-sulfation of glucosamine (GlcNAc) of the glycosaminoglycan in heparan sulfate. Modifies the GlcNAc-GlcA disaccharide repeating sugar backbone to make N-sulfated heparosan, a prerequisite substrate for later modifications in heparin biosynthesis. Has low deacetylase activity but high sulfotransferase activity.Specifications
NDST4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9H3R1 | |
NDST4 | |
KLH conjugated synthetic peptide between 185-212 amino acids from the N-terminal region of human NDST4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NDST4 | |
Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 4, Glucosaminyl N-deacetylase/N-sulfotransferase 4, NDST-4, N-heparan sulfate sulfotransferase 4, N-HSST 4, Heparan sulfate N-deacetylase 4, 3---, Heparan sulfate N-sulfotransferase 4, 282-, NDST4, HSST4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64579 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction