Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549497
Description
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Specifications
NDUFA1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O15239, O35683 | |
NDUFA1 | |
KLH conjugated synthetic peptide between 26-59 amino acids from the Central region of human NDUFA1. | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NDUFA1 | |
NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 1, Complex I-MWFE, CI-MWFE, NADH-ubiquinone oxidoreductase MWFE subunit, NDUFA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4694, 54405 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction