Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550462
Description
The antibody detects endogenous levels of total NDUFA2 protein.
NDUFA2 Polyclonal antibody specifically detects NDUFA2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
NDUFA2 | |
Polyclonal | |
Unconjugated | |
NDUFA2 | |
AV000592; B8; C1-B8; CCDC76; CD14; CIB8; CI-B8; complex I B8 subunit; complex I-B8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH:ubiquinone oxidoreductase subunit A2; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase subunit CI-B8; NDUFA2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17991, 4695 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O43678, Q9CQ75 | |
NDUFA2 | |
Full length fusion protein of Human NDUFA2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction