Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA7 Mouse anti-Human, Clone: OTI2F3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502640

Catalog No. 50-167-3678


Add to cart

Description

Description

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications

Specifications

NDUFA7
Monoclonal
0.63 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O95182
B14.5a, CI-B14.5a
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4701
Flow Cytometry, Western Blot
OTI2F3
Unconjugated
NDUFA7
Liquid
NDUFA7
Full length human recombit protein of human NDUFA7 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit