Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFA7 Mouse anti-Human, Clone: OTI2C3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525520

Catalog No. PIMA525520


Add to cart

Description

Description

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications

Specifications

NDUFA7
Monoclonal
0.9 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O95182
NADH dehydrogenase, ubiquinone 1 alpha subcomplex 7 14.5kDa, B14.5a, CI-B14.5a, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 7, NADH-ubiquinone oxidoreductase subunit B14.5a, complex I B14.5a subunit
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4701
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2C3
Unconjugated
NDUFA7
Liquid
NDUFA7
Full length human recombit protein of NDUFA7 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit