Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFAF2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550452

Catalog No. PA550452


Add to cart

Description

Description

The antibody detects endogenous levels of total NDUFAF2 protein.

Mimitin, a small mitochondrial protein, whose transcription is directly stimulated by c-Myc, is highly expressed in 80% of esophageal squamous cell carcinomas (ESCC). Suppression of Mimitin expression by RNA interference had no effect in cancerous cell lines such as human cervical carcinoma or hepatocarcinoma cell lines, but caused a decrease in cell proliferation in human glioblastoma, embryonic lung fibroblastic cells, and ESCC, suggesting Mimitin may play a special role in these types of cells. Mimitin expression is also regulated by MAPK kinases and IL-1, but not through the NF-kappa-B-related pathway. It will interact with the microtubular protein MAP1S and can affect the activities of caspase-3 and -7 in cells stimulated to develop apoptosis. Other experiments suggest that Mimitin also acts as a molecular chaperone for the assembly of the mitochondrial complex I.
Specifications

Specifications

NDUFAF2
Polyclonal
Unconjugated
NDUFAF2
Liquid
NDUFAF2
Fusion protein corresponding to a region derived from internal residues of human NADH dehydrogenase (ubiquinone) complex I, assembly factor 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
2.6 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8N183
NDUFA12L
Rabbit
IgG
100 μL
-20°C
Primary
91942
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit