Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFAF2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548555

Catalog No. PA548555

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Acts as a molecular chaperone for mitochondrial complex I assembly.
Specifications

Specifications

NDUFAF2
Polyclonal
PBS with 0.09% sodium azide
Q8N183
Mimitin, mitochondrial, B172-like, B172L, Myc-induced mitochondrial protein, MMTN, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2, NDUFA12-like protein, NDUFAF2, NDUFA12L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
91942
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NDUFAF2
Liquid
NDUFAF2
KLH conjugated synthetic peptide between 71-99 amino acids from the Central region of human NDUFAF2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit