Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFAF4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550713

Catalog No. PA550713


Add to cart

Description

Description

The antibody detects endogenous levels of total NDUFAF4 protein.

NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene are a cause of mitochondrial complex I deficiency.
Specifications

Specifications

NDUFAF4
Polyclonal
Unconjugated
NDUFAF4
Liquid
NDUFAF4
Synthetic peptide corresponding to residues near the C terminal of human NADH dehydrogenase (ubiquinone) complex I, assembly factor 4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
2.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9P032
C6orf66, HRPAP20
Rabbit
IgG
100 μL
-20°C
Primary
29078
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit