Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB10 Mouse anti-Human, Clone: OTI1H6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272915
Description
NDUFB10 Monoclonal specifically detects NDUFB10 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
NDUFB10 | |
Monoclonal | |
LYOPH | |
Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:150, CyTOF-ready | |
CI-PDSW, complex I PDSW subunit, Complex I-PDSW, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10 (22kD, PDSW), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10, NADH ubiquinone oxidoreductase PDSW subunit (RH 16p13.3), NADH-ubiquinone oxidoreductase PDSW subunit, PDSW | |
Mouse | |
20.6 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
OTI1H6 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
NDUFB10 | |
Full length human recombinant protein of human NDUFB10(NP_004539) produced in E.coli. | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
4716 | |
Human | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title