Learn More
NDUFB5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 238551AP
Catalog No. 50-173-4144
Add to cart
Description
Description
NDUFB5 is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
Specifications
NDUFB5
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43674, Q9CQH3
CI SGDH, Complex I SGDH, NDUFB5, SGDH
Rabbit
IgG
150 μL
-20°C
Primary
4711, 66046
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NDUFB5
Liquid
NDUFB5
NDUFB5 Fusion Protein Ag20907
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse