Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFB5 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen NDUFB5
Immunogen Synthetic peptides corresponding to NDUFB5(NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa) The peptide sequence was selected from the middle region of NDUFB5. Peptide sequence KLLRFYIALTGIPVAIFITLVNVFIGQAELAEIPEGYVPEHWEYYKHPIS
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15950020 View Documents Novus BiologicalsSupplier Diversity Partner
NBP15950020UL
20ul Each for $149.00
Add to cart
 
NBP159500 View Documents Novus BiologicalsSupplier Diversity Partner
NBP159500
100 ul Each for $329.00
Add to cart
 
Description

Description

NDUFB5 Polyclonal antibody specifically detects Antigen in Human, Rat samples. It is validated for Western Blotting.
Specifications

Specifications

NDUFB5
Rabbit
Polyclonal
Unconjugated
4711
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to NDUFB5(NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa) The peptide sequence was selected from the middle region of NDUFB5. Peptide sequence KLLRFYIALTGIPVAIFITLVNVFIGQAELAEIPEGYVPEHWEYYKHPIS
Primary
IgG
5 (16kD, SGDH), CISGDH, CI-SGDH, Complex I-SGDH, DKFZp686N02262, FLJ30597, MGC111204, MGC12314, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial, SGDH
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit