Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549342
Description
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Specifications
NDUFC2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O95298, Q9CQ54 | |
NDUFC2 | |
KLH conjugated synthetic peptide between 5-39 amino acids from the N-terminal region of human NDUFC2. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
NDUFC2 | |
NADH dehydrogenase [ubiquinone] 1 subunit C2, Complex I-B145b, CI-B145b, Human lung cancer oncogene 1 protein, HLC-1, NADH-ubiquinone oxidoreductase subunit B145b, NDUFC2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
293130, 4718, 68197 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction