Learn More
NDUFS2 Mouse anti-Human, Clone: OTI2G2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802325
Catalog No. 50-166-5943
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
NDUFS2 is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (complex I). Mammalian mitochondrial complex I is composed of at least 43 different subunits, 7 of which are encoded by the mitochondrial genome, and the rest are the products of nuclear genes. The iron-sulfur protein fraction of complex I is made up of 7 subunits, including this gene product. Complex I catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria.
Specifications
Specifications
NDUFS2
Monoclonal
Unconjugated
NDUFS2
Lyophilized
NDUFS2
Human recombit protein fragment corresponding to amino acids 205-463 of human NDUFS2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2G2
PBS with 8% trehalose and no preservative; pH 7.3
O75306
CI-49
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4720