Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFS2 Mouse anti-Human, Clone: OTI1D5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526536

Catalog No. PIMA526536


Add to cart

Description

Description

The protein encoded by this gene is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase. Mammalian mitochondrial complex I is composed of at least 43 different subunits, 7 of which are encoded by the mitochondrial genome, and the rest are the products of nuclear genes. The iron-sulfur protein fraction of complex I is made up of 7 subunits, including this gene product. Complex I catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

NDUFS2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O75306
CI-49, CI-49kD, NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial, NADH-ubiquinone oxidoreductase 49 kDa subunit, NADH-ubiquinone oxidoreductase NDUFS2 subunit, complex 1, mitochondrial respiratory chain, 49-KD subunit, complex I 49kDa subunit, complex I-49kD
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4720
Immunohistochemistry (Paraffin), Western Blot
OTI1D5
Unconjugated
NDUFS2
Liquid
NDUFS2
Human recombit protein fragment corresponding to amino acids 205-463 of NDUFS2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit