Learn More
Aviva Systems Biology Corp NDUFS3 Antibody - middle region (ARP56578_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDUFS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NDUFS3. Alternate names: CI-30ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61910