Learn More
NDUFV2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 153011AP
Catalog No. 50-173-0134
Add to cart
Description
Description
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
Specifications
NDUFV2
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P19234, P19404, Q9D6J6
NDUFV2
Rabbit
IgG
150 μL
-20°C
Primary
4729, 72900, 81728
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
NDUFV2
Liquid
NDUFV2
NDUFV2 Fusion Protein Ag7559
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat