Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFV3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549561

Catalog No. PA549561

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is one of at least forty-one subunits that make up the NADH-ubiquinone oxidoreductase complex. This complex is part of the mitochondrial respiratory chain and serves to catalyze the rotenone-sensitive oxidation of NADH and the reduction of ubiquinone. The encoded protein is one of three proteins found in the flavoprotein fraction of the complex. The specific function of the encoded protein is unknown. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].
Specifications

Specifications

NDUFV3
Polyclonal
PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P56181
NADH dehydrogenase [ubiquinone] flavoprotein 3, mitochondrial, Complex I-9kD, CI-9kD, NADH-ubiquinone oxidoreductase 9 kDa subunit, Renal carcinoma antigen NY-REN-4, NDUFV3
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4731
Immunohistochemistry, Western Blot
Unconjugated
NDUFV3
Liquid
NDUFV3
synthetic peptide corresponding to 35-62aa of human NDUV3
Affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit