Learn More
Cayman ChemicalSupplier Diversity Partner Nε-(1-Carboxymethyl)-L-lysine, 5746-04-3, 25 mg DFS Item
Manufacturer:  Cayman ChemicalSupplier Diversity Partner 1648325
An AGE, produced by the oxidative modification of glycated proteins during oxidative stress; increases with aging and during pathologies marked by oxidative stress; activates RAGE, triggering signaling through MAPKs and NF-κB
Catalog No. NC1464486