Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nebivolol hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Highly selective β1-adrenoceptor antagonist (Ki values are 0.88, 20, 44, 700, 1160, 2400 and 4000 nM at β1, 5-HT1A, β2, 5-HT2, α1, H1 and D2 receptors respectively). Induces vasodilation via a nitric oxide- and cGMP-dependent mechanism (EC50 = 11.36 μM in renal arteries) and displays antihypertensive activity in vivo. GRK/β -arrestin biased agonist at the β2 adrenoceptor.
Specifications
Quantity | 10 mg |
Formula Weight | 441.9 |
Percent Purity | >98% |
Chemical Name or Material | Nebivolol hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction