Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NECAB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565389
Description
Immunogen sequence: QDFHRALRCYV DFTGAQSHCL HVSAQKMLDG ASFTLYEFWQ DEASWRRHQQ SPGSKAFQRI LIDHLRAPDT LTTVFFPASW WIMNN Highest antigen sequence identity to the following orthologs - mouse 91%, rat 92%.
The protein encoded by this gene interacts with the amino-terminal domain of the neuron-specific X11-like protein (X11L), inhibits the association of X11L with amyloid precursor protein through a non-competitive mechanism, and abolishes the suppression of beta-amyloid production by X11L. This protein, together with X11L, may play an important role in the regulatory system of amyloid precursor protein metabolism and beta-amyloid generation. The protein is phosphorylated by NIMA-related expressed kinase 2, and localizes to the Golgi apparatus. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
NECAB3 | |
Polyclonal | |
Unconjugated | |
NECAB3 | |
2900010M17Rik; AI853434; amyloid beta (A4) precursor protein-binding family A member 1 binding protein; amyloid beta (A4) precursor protein-binding, family A, member 1 binding protein; amyloid beta (A4) precursor protein-binding, family A, member 2 binding protein; amyloid beta A4 protein-binding family A member 2-binding protein; Amyloid-beta A4 protein-binding family A member 2-binding protein; Apba2bp; dJ63M2.4; dJ63M2.5; EFCBP3; EF-hand calcium binding protein 3; mXB51; NECAB3; Nek2-interacting protein 1; neuronal calcium-binding protein 3; neuronal calcium-binding protein NECAB3; NIP1; N-terminal EF-hand calcium binding protein 3; N-terminal EF-hand calcium-binding protein 3; STIP3; synaptotagmin interacting protein 2; synaptotagmin interacting protein STIP3; SYTIP2; X11L-binding protein 51; XB51 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
63941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96P71 | |
NECAB3 | |
Recombinant Human NECAB3. Recombinant protein control fragment (Product #RP-106071). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction