Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NECAP1 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17958320UL

 View more versions of this product

Catalog No. NBP17958320


Add to cart

Description

Description

NECAP1 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

NECAP1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
NECAP1
Synthetic peptide directed towards the N terminal of human Necap1The immunogen for this antibody is Necap1. Peptide sequence RYFVIRIQDGTGRSAFIGIGFTDRGDAFDFNVSLQDHFKWVKQETEISKE.
30 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 1:1000
NP_080543
adaptin ear-binding coat-associated protein 1, adaptin-ear-binding coat-associated protein 1, DKFZP566B183, MGC131900, NECAP endocytosis associated 1, NECAP endocytosis-associated protein 1, NECAP-1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
25977
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit