Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Necdin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158244
Description
Necdin Polyclonal specifically detects Necdin in Human samples. It is validated for Western Blot.Specifications
Necdin | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q99608 | |
NDN | |
Synthetic peptides corresponding to NDN(necdin homolog (mouse)) The peptide sequence was selected from the middle region of NDN. Peptide sequence VALSNRMPMTGLLLMILSLIYVKGRGARESAVWNVLRILGLRPWKKHSTF. | |
100 μL | |
Apoptosis, Cancer, Tumor Suppressors | |
4692 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
HsT16328, necdin, necdin (mouse) homolog, necdin homolog (mouse), PWCR | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Rabbit: 100%; Mouse: 92%; Rat: 92%. | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Sheep | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title