Learn More
Mopec Non Ventilated Necropsy Table with Leg Frame
SDP
Integrally constructed large non vented sink with leg frame, which is creased toward the drain basin for complete and easy fluid drainage. Mopec™ Non Ventilated Necropsy Table with Leg Frame is economical in that it includes all the convenient features necessary to perform a necropsy.
Supplier:  Mopec OC500
Catalog No. 22-444-234
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
Description
Description
  • Stainless steel tabletop is 14 gauge stainless steel with #4 satin finish and built-in large integral sink
  • GFCI receptacles
  • Hydro aspirator that is designed to drain into sink, plumbed with water control valve, vacuum breaker and clear tubing
  • Hot and cold single lever mixing valve with swing spout faucet complete with vacuum breaker
  • Deck mounted chrome plated brass fixtures
  • Sink spray with high pressure hose
  • Four removable perforated grid plates
  • Grid pattern is perforated with 9.5mm diameter holes on 50.8mm centers
  • All plumbing and electrical work accomplished at factory
  • Only trade hook-up required
Warranty and Services
1 Year Warranty
Specifications
Specifications
Trimming Table
Trimming Tables
254 cm
GFCI Receptacles
94 cm
76.2 cm
Product Suggestions
Product Suggestions