Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Necrosulfonamide
Necroptosis inhibitor
$149.40 - $627.00
Specifications
Purity | >98% |
---|---|
Product Type | Necrosulfonamide |
Molecular Weight (g/mol) | 461.47 |
Inhibitors | Necrosulfonamide |
Description
Necroptosis inhibitor. Blocks mixed lineage kinase domain-like protein (MLKL), a critical substrate of receptor-interacting serine-threonine kinase 3 (RIP3) during necrosis. Prevents MLKL-RIP1-RIP3 necrosome complex from interacting with downstream necrosis effectors.Specifications
>98% | |
Necrosulfonamide | |
461.47 | |
Necrosulfonamide |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title