Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin-1/PVRL1 Mouse anti-Human, Clone: R1.302, Novus Biologicals™

Mouse Monoclonal Antibody

$228.85

Specifications

Antigen Nectin-1/PVRL1
Clone R1.302
Host Species Mouse
Isotype IgG1
Gene Alias CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1MGC16207, PVRR1nectin-1, SK-12
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25464301 View Documents Novus Biologicals
NBP2546430.1MG
0.1 mg Each for $228.85
Add to cart
 
Description

Description

Nectin-1/PVRL1 Monoclonal antibody specifically detects Nectin-1/PVRL1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Flow (Cell Surface), CyTOF-ready.
Specifications

Specifications

Nectin-1/PVRL1
Mouse
CD111, CD111 antigen, CLPED1ectodermal dysplasia 4 (Margarita Island type), ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig-like receptor, HIgRPVRR, HveC, HVECpoliovirus receptor-like 1, MGC142031, Nectin-1, OFC7nectin 1, poliovirus receptor-related 1 (herpesvirus entry mediator C), poliovirus receptor-related protein 1, PRR, PRR1MGC16207, PVRR1nectin-1, SK-12
CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL
R1.302
IgG1
Human
RUO
Affinity Purified
Primary
Phosphate buffered saline (PBS), pH 7.4 with 15mM Sodium Azide

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit