Learn More
Novus Biologicals™ Nectin-2/CD112 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2794315PLATES
 View more versions of this product
Catalog No. NB009902
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human Nectin-2/CD112.
Specifications
Specifications
HRP
This solid phase sandwich ELISA utilizes an antibody specific for Human Nectin-2/CD112.
15.625 pg/mL
5819
RUO
Nectin-2/CD112
5 Plates
15.625-1000 pg/mL
CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2
Sandwich ELISA
Antibody Pairs
Sandwich ELISA
  • Detection Antibody: 0.5 mg/mL rabbit anti-Nectin-2/CD112 polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.25 mg/mL of rabbit anti-Nectin-2/CD112 monoclonal antibody,
  • Standard: Each vial contains 50 ng of recombinant Nectin-2/CD112.
  • Storage is content dependent.
    Rabbit
    Human
    NECTIN2
    1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended.