Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nectin-2/CD112 Mouse anti-Rat, Clone: R2-525, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP1425290.05MG

Catalog No. NBP14252905


Add to cart

Description

Description

Nectin-2/CD112 Monoclonal antibody specifically detects Nectin-2/CD112 in Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Frozen),CyTOF.
Specifications

Specifications

Nectin-2/CD112
R2-525
Unconjugated
Phosphate buffered saline pH 7.4 with 0.09% sodium azide
NECTIN2
The antibody reacts with the CD112 antigen.
0.05 mg
Primary
5819
Rat
CyTOF, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
1.0 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
CD112, CD112 antigen, Herpes virus entry mediator B, Herpesvirus entry mediator B, herpesvirus entry protein B, HveB, HVEBpoliovirus receptor-like 2, nectin 2, Nectin-2, poliovirus receptor-related 2 (herpesvirus entry mediator B), poliovirus receptor-related protein 2, PRR2nectin-2, PVRR2poliovirus receptor related 2
Mouse
IgG1
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes CD112 Poliovirus Receptor Related 2 (PRR2), a new surface molecule, which has been reported to be homologous to the poliovirus receptor (CD155). Studies demonstrate that PRR2 expression is restricted to the myelo-monocytic and megakaryocytic lineages. PRR2 expression has also been detected on haematopoeitic progenitors expressing CD34.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit