Learn More
Tocris Bioscience™ Ned K Available on GSA/VA Contract for Federal Government customers only.
NAADP signaling inhibitor
$318.00 - $1339.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
59-761-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5976/10
Each for $318.00
Add to cart
59-765-0 View Documents Tocris Bioscience™
5976/50
Each for $1,339.00
Add to cart
Description
Description
NAADP inhibitor. Prevents Ca2+ oscillations in rat cardiomyocytes in vitro during simulated ischemia and reperfusion, but exhibits no significant effect on Ca2+ sparks. Significantly reduces infarct size in rat cardiac ischemia/reperfusion model in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.