Learn More
Sklar InstrumentsSupplier Diversity Partner Derf Needle Holder, Sklar, 4-3/4 in., Blunt, Straight, Serrated with Groove, Finger Rings, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 201547
Derf Needle Holder, Sklar, 4-3/4 in., Blunt, Straight, Serrated with Groove, Finger Rings, Stainless Steel. The Sklar Derf Needle Holder features short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures. Frequently utilized in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf Needle Holder to be easily controlled in smaller surgical areas. This product is straight with serrated tips, a central groove and a length of 4-3/4 inches.
Catalog No. 50-143-7455