Learn More
Sklar InstrumentsSupplier Diversity Partner TC Baumgartner Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218005
TC Baumgartner Needle Holder, Sklar, 5-1/4 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar TC Baumgartner Needle Holders feature heavy serrations which make this instrument capable of firmly holding larger needles and dense suture material. They are commonly used in procedures on the trunk and limbs. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with cross-serrated tips and a length of 5-1/4 inches.
Catalog No. 50-143-7695