Learn More
Sklar InstrumentsSupplier Diversity Partner TC Wangensteen Needle Holder, Sklar, 10-1/2 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 213510
TC Wangensteen Needle Holder, Sklar, 10-1/2 in., Blunt, Straight, Cross-Serrated, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's Tungsten Carbide Wangensteen Needle Holder has pyramid shaped teethto reduce grip slippage when using fine needles. The straight jaws are to be used with 1-0, 2-0 and 3-0 sutures. Sklar permanently bonds tungsten carbide inserts to the instrument, to ensure sharpness over a longer period of time, and maximize control and precision. TC instruments are identified by gold finger rings.
Catalog No. 50-143-7615