Learn More
Sklar InstrumentsSupplier Diversity Partner TC Castroviejo Needle Holder, Sklar, 5-1/2 in., Blunt/Blunt, Curved, Smooth, Thumb with Catch, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218048
TC Castroviejo Needle Holder, Sklar, 5-1/2 in., Blunt/Blunt, Curved, Smooth, Thumb with Catch, Tungsten Carbide / Stainless Steel. Sklar Castroviejo Tungsten Carbide Needle Holder is commonly used for suturing in ophthalmologic procedures. It may also be used in other microsurgical procedures. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles. This product is curved with smooth tips and a length of 5-1/2 inches. It also features a locking mechanism.
Catalog No. 50-143-7734