Learn More
Sklar InstrumentsSupplier Diversity Partner TC Jacobson Needle Holder, Sklar, 8-1/4 in., Blunt, Curved, Smooth, Thumb, Tungsten Carbide/ Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218258
TC Jacobson Needle Holder, Sklar, 8-1/4 in., Blunt, Curved, Smooth, Thumb, Tungsten Carbide / Stainless Steel. Sklar's Jacobson Tungsten Carbide Microvascular Needle Holder is primarily used for suturing in microvascular and cardiovascular procedures. It may also be known as a Jacobson-Castroviejo Needle Holder. The finely cross-serrated, tapered jaws are able to securely hold very fine sutures. Serrations on TC instruments are designed to stay sharp for a significantly longer period of time than standard instruments. Sklar also permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is curved with micro-profile, cross-serrated jaws and an overall length of 8-1/4 inches. There is no locking mechanism.
Catalog No. 50-143-7822