Learn More
Organomation Set of 12 4in. X 19ga. Stainless Steel Needles with a Plastic Hub
SDP
Supplier:  Organomation NA060419
Encompass_Preferred
Set of one dozen luer lock needles. These needles are 4 inch by 19 gauge, with a 304 stainless steel shaft and a polypropylene hub.
Catalog No. NC0593680