Neese™ ComforTuff FR™ Work Shirts

Manufacturer: Neese Industries Inc. VU7SHYLL

 View all options for this product

Catalog No. 19-043-237

  Check Availability
Add to Cart


    Arc Flash Rating 8.7 Calories/cm2
    Certifications/Compliance NFPA 70E™, 1971, 1977, NFPA 2112 and ASTM F1506
    Chest Size 42 to 44 in.
    Color Yellow
    Material Cotton, Nylon
    Size Large
    Hazard Rating HRC 2
    Length (English) Sleeve 34 in.
    Length (Metric) Sleeve 86.36cm
    Size (Metric) Neck 40.64 to 41.91cm

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit