Neese Industries Acid Suit 96 Coat

Manufacturer: Neese Industries Inc. 96 SC2X

 View more versions of this product

Catalog No. 19-046-103D

  Check Availability
Add to Cart

    Specifications

    Size 2X-Large
    Includes Storm flap, raglan shoulder
    Material PVC, Polyester
    Closure Type Front snap
    Hood Type Snap
    Length (English) 48 in.
    Length (Metric) 121.92cm
    Resistance Flame
    Collar Type Stand up
    Color Green
    Construction PVC, Polyester Knit
    For Use With (Application) Chemical, rain, flame

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit