Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neese Acid Suit 96 Jackets
SDP

Green PVC/polyester knit jackets in a variety of styles

$111.00 - $755.00

Specifications

Material PVC, Polyester
Color Green
Closure Type Snap
Construction PVC, Polyester Knit
Resistance Flame
View More Specs

Products 5
Catalog Number Mfr. No. Size Seam Type Price Quantity & Availability  
Catalog Number Mfr. No. Size Seam Type Price Quantity & Availability  
19-020316
SDP
View Documents
Neese
96001001GRNM
Medium Welded
Each for $111.00
Add to cart
 
19-069215
SDP
View Documents
Neese
96001011GRNM
Medium Double-welded
Each for $755.00
Add to cart
 
19-046-148A
SDP
View Documents
Neese
96JS
Small Double-welded This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-042902
SDP
View Documents
Neese
96JL
Large Double-welded This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
19-069216
SDP
View Documents
Neese
96SJGRNXLG
X-Large Double-welded This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Snap front
  • Storm flap
  • Available with attached hood with drawstring or snaps for hood
  • 19-046-148A and 19-042-902 have double-welded seams

Compliance

ASTM D6413: Flame Resistance and ASTM F903: Resistance To Penetration By Liquids

Specifications

Specifications

PVC, Polyester
Snap
Flame
76.2 cm
Green
PVC, Polyester Knit
30 in.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit