Neese Industries PVC Coveralls

Manufacturer: Neese Industries Inc. 96ACAM

 View more versions of this product

Catalog No. 19-069-227

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM D6413, ASTM F903
    Material PVC, Polyester
    Includes Attached hood, zipper front, elastic wrist, ankle and face openings
    Seam Type Double welded
    Color Green
    Size Medium
    For Use With (Application) Sewer works, containment tank cleaning, incidental chemical splash and general industrial applications
    Closure Type Snap Front, Zipper
    Weight (English) 0.81 lb.
    Weight (Metric) 0.36kg

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit