Neese™ Three-Piece Acid Suit
Offers splash protection with a broad range of chemicals
Manufacturer: Neese Industries Inc. I96SMD
 View more versions of this product
Catalog No. 19-168-352B
  Check Availability
Add to Cart
    Specifications
    Cuff Style Elastic
    Color Green
    Material Polyester/PVC
    Size Medium
    Hood Type Detached
    • Constructed of heavyweight 13 oz. (368g) PVC/polyester knit coated on both sides
    • Jacket is a minimum of 30 in. long (76cm) with adjustments for various sizes
    • Stand-up collar with snaps to allow for detachable hood
    • Double welded seams
    • Storm flap conceals dielectric snaps
    • Flame retardant
  • Description & Specifications
Related Products
Product Content Feedback