Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neese Neese™ Three-Piece I36S Rainsuit

Economical chemical-splash protection for a variety of industrial applications

Manufacturer:  Neese I36SXL

 View more versions of this product

Catalog No. 19-168-353D


Add to cart

Description

Description

  • Made 32mm PVC/nylon/PVC with electronically welded seams
  • Jacket has snap-on hood, storm flap, and nonconductive snaps
  • Bib-style, chest-high trousers feature safety fly and take-up snaps on cuffs and waist

Specifications

Specifications

Detached
124.4cm
Nylon/PVC
49 in.
Yellow
X-Large
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit