Learn More
Neese Universal 35 Rainwear Coats
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Hood Type:
Attached
Detachable
Description
- PVC / Nylon / PVC
- Flame Resistant (ASTM D6413)
- Coat style that is minimum of 48 in. long with adjustments for various sizes
- Snap front
- Storm flap
- Attached hood with drawstring closure or stand up collar with snaps for hood
- Raglan shoulder design to allow for easy movement
Specifications
Size | X-Large |
Material | Nylon, PVC |
Color | Yellow |
Closure Type | Snap, Storm Flap |
Hood Type | Attached |
Sleeve Style | Raglan |