Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nefazodone hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
82752-99-6
C25H33Cl2N5O2
506.47
MFCD00935760
DYCKFEBIOUQECE-UHFFFAOYSA-N
nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
54911
CHEBI:7495
hydrogen 1-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-3-ethyl-4-(2-phenoxyethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one chloride
[H+].[Cl-].CCC1=NN(CCCN2CCN(CC2)C2=CC=CC(Cl)=C2)C(=O)N1CCOC1=CC=CC=C1
This item is not returnable. View return policy
82752-99-6
C25H33Cl2N5O2
506.47
MFCD00935760
DYCKFEBIOUQECE-UHFFFAOYSA-N
nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
54911
CHEBI:7495
hydrogen 1-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-3-ethyl-4-(2-phenoxyethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one chloride
[H+].[Cl-].CCC1=NN(CCCN2CCN(CC2)C2=CC=CC(Cl)=C2)C(=O)N1CCOC1=CC=CC=C1

5-HT2A antagonist and 5-HT uptake inhibitor. Antidepressant

Serotonin 5-HT2A receptor antagonist (Ki = 5.8 nM) and inhibitor of serotonin and noradrenalin uptake (IC50 values are 290 and 300 nM respectively). Displays no activity at 5-HT1B and 5-HT1D receptors. Active in models predictive of antidepressant potential.
Quantity 10 mg
Formula Weight 506.47
Percent Purity >99%
Chemical Name or Material Nefazodone hydrochloride

Recommended Storage : Desiccate at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit