Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nefazodone hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
82752-99-6
C25H33Cl2N5O2
506.47
MFCD00935760
DYCKFEBIOUQECE-UHFFFAOYSA-N
nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
54911
CHEBI:7495
hydrogen 1-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-3-ethyl-4-(2-phenoxyethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one chloride
[H+].[Cl-].CCC1=NN(CCCN2CCN(CC2)C2=CC=CC(Cl)=C2)C(=O)N1CCOC1=CC=CC=C1
This item is not returnable. View return policy
82752-99-6
C25H33Cl2N5O2
506.47
MFCD00935760
DYCKFEBIOUQECE-UHFFFAOYSA-N
nefazodone hydrochloride, nefazodone hcl, serzone, dutonin, menfazona, rulivan, reseril, 1-3-4-3-chlorophenyl piperazin-1-yl propyl-3-ethyl-4-2-phenoxyethyl-1h-1,2,4-triazol-5 4h-one hydrochloride, nefadar
54911
CHEBI:7495
hydrogen 1-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-3-ethyl-4-(2-phenoxyethyl)-4,5-dihydro-1H-1,2,4-triazol-5-one chloride
[H+].[Cl-].CCC1=NN(CCCN2CCN(CC2)C2=CC=CC(Cl)=C2)C(=O)N1CCOC1=CC=CC=C1

5-HT2A antagonist and 5-HT uptake inhibitor. Antidepressant

Serotonin 5-HT2A receptor antagonist (Ki = 5.8 nM) and inhibitor of serotonin and noradrenalin uptake (IC50 values are 290 and 300 nM respectively). Displays no activity at 5-HT1B and 5-HT1D receptors. Active in models predictive of antidepressant potential.
Quantity 50 mg
Formula Weight 506.47
Percent Purity >99%
Chemical Name or Material Nefazodone hydrochloride

Recommended Storage : Desiccate at Room Temperature