Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nefazodone hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Serotonin 5-HT2A receptor antagonist (Ki = 5.8 nM) and inhibitor of serotonin and noradrenalin uptake (IC50 values are 290 and 300 nM respectively). Displays no activity at 5-HT1B and 5-HT1D receptors. Active in models predictive of antidepressant potential.
Specifications
Quantity | 10 mg |
Formula Weight | 506.47 |
Percent Purity | >99% |
Chemical Name or Material | Nefazodone hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction