Learn More
NEFH Mouse anti-Human, Clone: RNF404, Abnova™
Mouse monoclonal antibody raised against human NEFH.
Manufacturer:  Abnova Corporation MAB12678
Catalog No. 89-934-673
Add to cart
Description
Description
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the heavy neurofilament protein. This protein is commonly used as a biomarker of neuronal damage and susceptibility to amyotrophic lateral sclerosis (ALS) has been associated with mutations in this gene. [provided by RefSeq]
Specifications
Specifications
NEFH
RNF404
Mouse monoclonal antibody raised against human NEFH.
In PBS (0.09% sodium azide).
NFH
Mouse
IgG2a
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Frozen sections) (1:50-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:100-1:500) The optimal working dilution should be determined by the end user.
Liquid
NEFH
Human NEFH
Protein G purification
RUO
Primary
4744