Learn More
NEFH (phospho KSP/KEP) Mouse anti-Human, Pig, Rat, Clone: NE14, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB14382
Catalog No. 89-034-464
Add to cart
Description
Description
Mouse monoclonal antibody raised against native pig NEFH.
  • High specificity
  • High reactivity
Specifications
Specifications
NEFH (phospho KSP/KEP)
NE14
Mouse monoclonal antibody raised against native pig NEFH.
In 10 mM PBS.
Mouse
IgG1
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Pig, Rat
Flow Cytometry, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Crude neurofilament preparation from porcine spinal cord.
Protein A/G purification
RUO
Primary
This monoclonal antibody identified as heavy subunit of neurofilaments (NEFH). It reacts specifically with the phosphorylated KSP/KEP segment at the C-terminus of the heavy subunit of neurofilaments. After dephosphorylation of neurofilaments with alkaline phosphatase, this antibody no longer binds.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)