Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Negative Control for Mouse IgG2b Ab-1 with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Mouse IgG2b Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Negative Control for Mouse IgG2b Ab-1
Clone NCG2B.01
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
N1391P0
View Documents
Epredia™
NC1391P0
Concentrated 100 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
N1391P1ABX
View Documents
Epredia™
NC1391P1ABX
Concentrated 100 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
N1391PABX
View Documents
Epredia™
NC1391PABX
Concentrated 200 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
N1391P
View Documents
Epredia™
NC1391P
Concentrated 1 mL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

This mouse monoclonal IgG2b was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG2b does not react with any antigen of human or animal cells. The monoclonal IgG2b was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG2b should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG2b antibodies.

Host Species: Mouse

Clone: NCG2B.01

Isotype: IgG2b/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Negative Control for Mouse IgG2b Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
All species
Primary
NCG2B.01
Monoclonal
Mouse
Negative Controls
IgG2b κ
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit