Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Negative Control for Rabbit Monoclonal Rabbit anti-Human, All Species, PE/Cy5.5, Clone: NCRBM/1520R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254471PECY55

Catalog No. N254471PEC5


Add to cart

Description

Description

Negative Control for Rabbit Monoclonal Monoclonal antibody specifically detects Negative Control for Rabbit Monoclonal in Human, All Species samples. It is validated for Flow Cytometry.
Specifications

Specifications

Negative Control for Rabbit Monoclonal
NCRBM/1520R
Flow Cytometry
Conjugated, Purified
Recombinant full-length human KRT7 protein
Protein A purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
All, Human
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Rabbit
IgG Kappa
0.1 mL
Primary
To serve as a true negative control for various procedures, this rabbit monoclonal IgG should be used under the identical conditions and at the same dilution as the rabbit monoclonal antibody being used for a positive reaction.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit